Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IGFBP-4 Antibody (IBP180), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP232771F
Description
IGFBP-4 Monoclonal antibody specifically detects IGFBP-4 in Human samples. It is validated for ELISA.Specifications
IGFBP-4 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
IGFBP4 | |
Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with recombinant human IGFBP-4 expressed in mammalian cell line. | |
0.2 mL | |
Primary | |
Human N-terminal IGFBP-4 fragment (NT-IGFBP-4) | |
Store at 4C in the dark. | |
IgG2a |
ELISA | |
IBP180 | |
ELISA, Sandwich ELISA | |
BP-4, HT29-IGFBP, IBP4insulin-like growth factor-binding protein 4, IGF-binding protein 4, IGFBP-4IBP-4, insulin-like growth factor binding protein 4 | |
Mouse | |
Protein A purified | |
RUO | |
3487 | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction