Learn More
IGFBP-5 Antibody (MM0344-8D21), Alexa Fluor™ 488, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP212366AF488
Catalog No. NB212366488
Only null left
Add to Cart
Description
Description
IGFBP-5 Monoclonal specifically detects IGFBP-5 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
IGFBP-5
Monoclonal
Alexa Fluor 488
50mM Sodium Borate with 0.05% Sodium Azide
IGFBP5
Human recombinant IGFBP-5
0.1 mL
3488
Store at 4C in the dark.
IgG2
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
MM0344-8D21
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
IBP5IBP-5, IGF-binding protein 5, IGFBP-5, insulin-like growth factor binding protein 5, insulin-like growth factor-binding protein 5
Mouse
Protein G purified
Primary
Human
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.