Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IGFBP-5 Antibody (RM0097-12C23), Alexa Fluor™ 350, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP122453AF350
Description
IGFBP-5 Monoclonal specifically detects IGFBP-5 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
IGFBP-5 | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
IGFBP5 | |
Mouse recombinant protein of IGFBP-5 | |
0.1 mL | |
3488.0 | |
Mouse | |
IgG2 |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
RM0097-12C23 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
IBP5IBP-5, IGF-binding protein 5, IGFBP-5, insulin-like growth factor binding protein 5, insulin-like growth factor-binding protein 5 | |
Rat | |
Protein A or G purified | |
Primary | |
No cross activity to other mouse IGFBPs | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction