Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IGFBP-6 Antibody (104), Alexa Fluor™ 350

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290597AF350
Description
IGFBP-6 Monoclonal specifically detects IGFBP-6 in Mouse samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
IGFBP-6 | |
Monoclonal | |
Alexa Fluor 350 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
104 | |
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
IBP6, IBP-6, IGF binding protein 6, IGF-binding protein 6, IGFBP-6, insulin-like growth factor binding protein 6, insulin-like growth factor-binding protein 6 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse IGFBP-6 (NP_032370.2; Met1-Gly238). | |
0.1 mL | |
Cell Cycle and Replication | |
3489 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction