Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IGFBP-L1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP31548720UL
Description
IGFBP-L1 Polyclonal antibody specifically detects IGFBP-L1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
IGFBP-L1 | |
Polyclonal | |
PBS, 50% glycerol, pH7.3 | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
Unconjugated | |
bA113O24.1, IGFBPL1 insulin-like growth factor binding protein-like 1, IGFBP-RP4 | |
A synthetic peptide corresponding to a sequence within amino acids 150-250 of human IGFBP-L1 (NP_001007564.1). | |
20 μg | |
Cell Biology, Signal Transduction | |
347252 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction