Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IGFBP-rp1/IGFBP-7 Antibody (003), DyLight 405

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290174V
Description
IGFBP-rp1/IGFBP-7 Monoclonal specifically detects IGFBP-rp1/IGFBP-7 in Human samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.Specifications
IGFBP-rp1/IGFBP-7 | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
No cross-reactivity in ELISA with: Mouse IGFBP6 Human IGFBP4 | |
Store at 4°C in the dark. | |
IgG |
ELISA, Immunofluorescence | |
3 | |
ELISA, Immunocytochemistry/Immunofluorescence | |
angiomodulin, FSTL2, IBP-7, IGFBP-7PGI2-stimulating factor, IGFBP-7v, IGFBP-rP1, IGFBPRP1, insulin-like growth factor binding protein 7, insulin-like growth factor-binding protein 7, MAC25 protein, MAC25IGF-binding protein 7, Prostacyclin-stimulating factor, PSFAGM, TAF, Tumor-derived adhesion factor | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human IGFBP-rp1/IGFBP-7 (Q16270; Met1-Leu282). | |
0.1 mL | |
Biologically Active Proteins | |
3490 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction