Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IGFBP-rp1/IGFBP-7 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 3 publications
$382.00 - $610.00
Specifications
Antigen | IGFBP-rp1/IGFBP-7 |
---|---|
Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
IGFBP-rp1/IGFBP-7 Polyclonal specifically detects IGFBP-rp1/IGFBP-7 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
IGFBP-rp1/IGFBP-7 | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
3490 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:EQGPSIVTPPKDIWNVTGAQVYLSCEVIGIPTPVLIWNKVKRGHYGVQRTELLPGDRDNLAIQTRGGPEKHEVTGWVLVSPLSKEDAGEYECHASNSQGQA | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
Polyclonal | |
Rabbit | |
Human | |
angiomodulin, FSTL2, IBP-7, IGFBP-7PGI2-stimulating factor, IGFBP-7v, IGFBP-rP1, IGFBPRP1, insulin-like growth factor binding protein 7, insulin-like growth factor-binding protein 7, MAC25 protein, MAC25IGF-binding protein 7, Prostacyclin-stimulating factor, PSFAGM, TAF, Tumor-derived adhesion factor | |
IGFBP7 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title