Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IGFBP-rp1/IGFBP-7 Rabbit anti-Human, Clone: 65, Novus Biologicals™

Rabbit Monoclonal Antibody
$342.00 - $524.00
Specifications
Antigen | IGFBP-rp1/IGFBP-7 |
---|---|
Clone | 65 |
Dilution | Western Blot 1:500-1:1000, Immunoprecipitation 1-2 ul/mg of lysate |
Applications | Western Blot, Immunoprecipitation |
Classification | Monoclonal |
Description
IGFBP-rp1/IGFBP-7 Monoclonal specifically detects IGFBP-rp1/IGFBP-7 in Human samples. It is validated for Immunoprecipitation.Specifications
IGFBP-rp1/IGFBP-7 | |
Western Blot 1:500-1:1000, Immunoprecipitation 1-2 ul/mg of lysate | |
Monoclonal | |
Purified | |
RUO | |
Human | |
angiomodulin, FSTL2, IBP-7, IGFBP-7PGI2-stimulating factor, IGFBP-7v, IGFBP-rP1, IGFBPRP1, insulin-like growth factor binding protein 7, insulin-like growth factor-binding protein 7, MAC25 protein, MAC25IGF-binding protein 7, Prostacyclin-stimulating factor, PSFAGM, TAF, Tumor-derived adhesion factor | |
Recombinant Human IGFBP-rp1/IGFBP-7 Protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
65 | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Biologically Active Proteins | |
0.2 um filtered solution in PBS | |
3490 | |
IgG | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title