Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IGFBP2 Monoclonal Antibody (04)
GREENER_CHOICE

Catalog No. PIMA529854
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA529854 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA529854 Supplier Invitrogen™ Supplier No. MA529854
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Rhesus IGFBP-2/IGFBP2.

IGFBP2 (insulin-like growth factor binding protein 2) is a member of the ISGBP family which bind various IGFs. IGFBP2 is overexpressed in a wide spectrum of cancers, including glioma, prostate cancer, synovial sarcoma, neuroblastoma, colon cancer, adrenocortical cancer, lung cancer, Wilms' tumor, and hepatoblastoma. The overexpression of IGFBP2 also correlates with the aggressiveness of some tumors. IGFBP2 activates the expression of matrix metalloprotease 2, which contributes to cell invasiveness.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IGFBP2
Applications ELISA
Classification Monoclonal
Clone 04
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene IGFBP2
Gene Accession No. 0
Gene Alias AI255832; BP2; BRL-BP; IBP2; IBP-2; IGF binding protein 2; IGF-binding protein 2; IGFBP; IGFBP2; IGFBP-2; IGF-BP53; ILGFBPA; Insulin like growth factor binding protein; insulin like growth factor binding protein 2; insulin-like growth factor binding protein 2; insulin-like growth factor binding protein 2, 36kDa; insulin-like growth factor-binding protein 2; mIGFBP-2; OTTHUMP00000207753
Gene Symbols IGFBP2
Host Species Mouse
Immunogen Recombinant Rhesus IGFBP-2/IGFBP2 protein.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 696214
Target Species Rhesus Macaque
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.