Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ IGFBP2 Recombinant Rabbit Monoclonal Antibody (7G11)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA549890
Description
IGFBP2 Recombinant Monoclonal Antibody for Western Blot, IHC (P), ELISA
IGFBP2 (insulin-like growth factor binding protein 2) is a member of the ISGBP family which bind various IGFs. IGFBP2 is overexpressed in a wide spectrum of cancers, including glioma, prostate cancer, synovial sarcoma, neuroblastoma, colon cancer, adrenocortical cancer, lung cancer, Wilms' tumor, and hepatoblastoma. The overexpression of IGFBP2 also correlates with the aggressiveness of some tumors. IGFBP2 activates the expression of matrix metalloprotease 2, which contributes to cell invasiveness.
Specifications
IGFBP2 | |
Recombinant Monoclonal | |
0.48 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P18065 | |
IGFBP2 | |
A synthesized peptide derived from human IGFBP2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
7G11 | |
Unconjugated | |
IGFBP2 | |
AI255832; BP2; BRL-BP; IBP2; IBP-2; IGF binding protein 2; IGF-binding protein 2; IGFBP; IGFBP2; IGFBP-2; IGF-BP53; ILGFBPA; Insulin like growth factor binding protein; insulin like growth factor binding protein 2; insulin-like growth factor binding protein 2; insulin-like growth factor binding protein 2, 36kDa; insulin-like growth factor-binding protein 2; mIGFBP-2; OTTHUMP00000207753 | |
Rabbit | |
Affinity chromatography | |
RUO | |
3485 | |
-20°C or -80°C if preferred | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction