Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IGFBP3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA579451
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA579451 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA579451 Supplier Invitrogen™ Supplier No. PA579451
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: rat heart tissue, rat brain tissue, rat liver tissue, rat PC-12 whole cellhuman U-87MG whole cellmouse kidney tissue, mouse heart tissue, mouse brain tissue. IHC: Human Intestinal Cancer tissue.

IGFBP3 is a gene that belongs to the insulin-like growth factor binding protein (IGFBP) family. The gene encodes a protein that contains both an IGFBP domain and a thyroglobulin type-I domain. This protein is known to form a ternary complex with insulin-like growth factor acid-labile subunit (IGFALS) and either insulin-like growth factor (IGF) I or II. The complex circulates in the plasma and prolongs the half-life of IGFs while also altering their interaction with cell surface receptors. Various isoforms of the gene have been identified through alternate transcriptional splice variants. Diseases associated with IGFBP3 include Insulin-Like Growth Factor I and Acid-Labile Subunit Deficiency.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IGFBP3
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide
Gene IGFBP3
Gene Accession No. P15473, P17936, P47878
Gene Alias acid stable subunit of the 140 K IGF complex; AI649005; binding protein 29; binding protein 53; BP53; BP-53; Growth hormone dependent binding protein; growth hormone-dependent binding protein; I79_011450; IBP3; IBP-3; IGF binding protein 3; IGF-binding protein 3; IGFBP; Igfbp3; IGF-BP3; IGFBP-3; IGgfbp3; Insulin like growth factor binding protein; insulin like growth factor binding protein 3; insulin-like growth factor binding protein 3; Insulin-like growth factor binding protein 3 precursor; insulin-like growth factor-binding protein (IGF-BP3); insulin-like growth factor-binding protein 3; tcag7.703
Gene Symbols IGFBP3
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence at the C-terminus of human IGFBP-3 (214-252aa RREMEDTLNHLKFLNVLSPRGVHIPNCDKKGFYKKKQCR).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16009, 24484, 3486
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.