Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGFBP3 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983253
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3253 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3253 Supplier Bioss Supplier No. BS1434R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Insulin like growth factor binding protein 3 (IGFBP3) is a member of the superfamily of insulin like growth factor (IGF) binding proteins which include six high affinity IGF binding proteins (IGFBP) and at least four low affinity binding proteins referred to as IGFBP related proteins (IGFBPrP). The IGFBP members are cysteine rich proteins with conserved cysteine residues clustered in the amino terminal and the carboxy terminal regions of the molecule. The cDNA sequence encoding the mature human IGFBP3 is fused to the signal peptide of CD33. The 265 amino acid residue recombinant mature human IGFBP3 has a calculated molecular mass of approximately 29 kDa. Due to glycosylation, the recombinant protein migrates as 41 kDa in SDS-PAGE.

Specifications

Antigen IGFBP3
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene IGFBP3
Gene Accession No. P17936
Gene Alias acid stable subunit of the 140 K IGF complex; AI649005; binding protein 29; binding protein 53; BP-53; growth hormone-dependent binding protein; I79_011450; IBP3; IBP-3; IGF binding protein 3; IGF-binding protein 3; IGFBP; IGFBP3; IGF-BP3; Igfbp-3; IGgfbp3; Insulin like growth factor binding protein; insulin like growth factor binding protein 3; insulin-like growth factor binding protein 3; insulin-like growth factor-binding protein (IGF-BP3); Insulin-like growth factor-binding protein 3; tcag7.703
Gene Symbols IGFBP3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human IGFBP3.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3486
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.