Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IGFBP4 Monoclonal Antibody (C15)
GREENER_CHOICE

Catalog No. PIMA541608
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA541608 200 μL
PIMA543749 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA541608 Supplier Invitrogen™ Supplier No. MA541608
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

IGFBP4 Monoclonal Antibody for Western Blot

This gene is a member of the insulin-like growth factor binding protein family and encodes a protein with an IGFBP domain and a thyroglobulin type-I domain. The protein binds both insulin-like growth factors I and II and circulates in the plasma in both glycosylated and non-glycosylated forms. Binding of this protein prolongs the half-life of the IGFs and alters their interaction with cell surface receptors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IGFBP4
Applications Western Blot
Classification Monoclonal
Clone C15
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene IGFBP4
Gene Accession No. P21744, P22692, P47879
Gene Alias AI875747; Band 3A; BP-4; Deb2; differentially expressed in B16F10 2; HT29-IGFBP; IBP4; IBP-4; IGF-binding protein 4; IGFBP; Igfbp4; IGF-BP4; IGFBP-4; Insulin like growth factor binding protein; insulin like growth factor binding protein 4; insulin-like growth factor binding protein 4; insulin-like growth factor binding protein-4; insulin-like growth factor-binding protein 4; insulin-like growth factor-binding protein-4; LOW QUALITY PROTEIN: insulin-like growth factor-binding protein 4
Gene Symbols IGFBP4
Host Species Mouse
Immunogen Recombinant protein Insulin Like Growth Factor Binding Protein 4. The antigen corresponds to amino acid range 22-258 of the target protein.
Purification Method Protein A/G
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16010, 3487, 360622
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.