Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IGFBP4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115390
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115390 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115390 Supplier Invitrogen™ Supplier No. PA5115390
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total IBP4.

This gene is a member of the insulin-like growth factor binding protein family and encodes a protein with an IGFBP domain and a thyroglobulin type-I domain. The protein binds both insulin-like growth factors I and II and circulates in the plasma in both glycosylated and non-glycosylated forms. Binding of this protein prolongs the half-life of the IGFs and alters their interaction with cell surface receptors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IGFBP4
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene IGFBP4
Gene Accession No. P22692
Gene Alias AI875747; Band 3A; BP-4; Deb2; differentially expressed in B16F10 2; HT29-IGFBP; IBP4; IBP-4; IGF-binding protein 4; IGFBP; Igfbp4; IGF-BP4; IGFBP-4; Insulin like growth factor binding protein; insulin like growth factor binding protein 4; insulin-like growth factor binding protein 4; insulin-like growth factor binding protein-4; insulin-like growth factor-binding protein 4; insulin-like growth factor-binding protein-4; LOW QUALITY PROTEIN: insulin-like growth factor-binding protein 4
Gene Symbols IGFBP4
Host Species Rabbit
Immunogen A synthesized peptide derived from human IGFBP4(Accession P22692), corresponding to amino acid residues S190-G240.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3487
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.