Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IGFBP6 Monoclonal Antibody (08)
GREENER_CHOICE

Catalog No. PIMA530518
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA530518 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA530518 Supplier Invitrogen™ Supplier No. MA530518
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human IGFBP6/IBP6.

IGF-BPs control the distribution, function and activity of IGFs in various cell tissues and body fluids. IGF-BP6, which specifically inhibits IGF-II actions, is produced by bone cells and is the major IGF-BP present in cerebrospinal fluid. IGF-BP6 has been shown to inhibit IGF-II-dependent cancers such as neuroblastoma, colon cancer and rhabdomyosarcoma.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IGFBP6
Applications ELISA
Classification Monoclonal
Clone 08
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene IGFBP6
Gene Accession No. P24592
Gene Alias IBP6; IBP-6; IGF binding protein 6; IGF-binding protein 6; IGFBP; IGFBP6; IGF-BP6; Igfbp-6; Insulin like growth factor binding protein; insulin like growth factor binding protein 6; insulin-like growth factor binding protein 6; INSULIN-LIKE GROWTH FACTOR BINDING PROTEIN 6 PRECURSOR (IGFBP-6) (IBP-6) (IGF-BINDING PROTEIN 6); insulin-like growth factor-binding protein 6; RBP6A
Gene Symbols IGFBP6
Host Species Mouse
Immunogen Recombinant Human IGFBP6/IBP6 Protein (Met1-Gly240).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3489
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.