Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGFBP6 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501985875
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-5875 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-5875 Supplier Bioss Supplier No. BS4064R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IGFBP6 encodes for a 216-amino acid protein with a calculated molecular weight of 22,847. The protein was found to be an autocrine growth inhibitor. Recombinant IGFBP6 was shown to inhibit growth of HaCat cells and other keratinocyte cell lines.

Specifications

Antigen IGFBP6
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene IGFBP6
Gene Accession No. P24592
Gene Alias IBP6; IBP-6; IGF binding protein 6; IGF-binding protein 6; IGFBP; IGFBP6; Igfbp-6; Insulin like growth factor binding protein; insulin like growth factor binding protein 6; insulin-like growth factor binding protein 6; INSULIN-LIKE GROWTH FACTOR BINDING PROTEIN 6 PRECURSOR (IGFBP-6) (IBP-6) (IGF-BINDING PROTEIN 6); Insulin-like growth factor-binding protein 6
Gene Symbols IGFBP6
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human IGFBP6.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3489
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.