Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PeproTech IGFBP7 Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™

Catalog No. 502718587
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 μg
50 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-271-8587 500 μg
500P234BT25 25 μg
500P234BT50 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 50-271-8587 Supplier PeproTech Supplier No. 500P234BT500UG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

AA Sequence of recombinant protein: SSSDTCGPCE PASCPPLPPL GCLLGETRDA CGCCPMCARG EGEPCGGGGA GRGYCAPGME CVKSRKRRKG KAGAAAGGPG VSGVCVCKSR YPVCGSDGTT YPSGCQLRAA SQRAESRGEK AITQVSKGTC EQGPSIVTPP KDIWNVTGAQ VYLSCEVIGI PTPVLIWNKV KRGHYGVQRT ELLPGDRDNL AIQTRGGPEK HEVTGWVLVS PLSKEDAGEY ECHASNSQGQ ASASAKITVV DALHEIPVKK GEGAEL. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human IGF-BP7. Anti-Human IGF-BP7-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect hIGF-BP7 by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human IGF-BP7 (500-P234) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant hIGF-BP7. Western Blot: To detect hIGF-BP7 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hIGF-BP7 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P234BT-1MG will be provided as 2 x 500 μg

This gene encodes a member of the insulin-like growth factor (IGF)-binding protein (IGFBP) family. IGFBPs bind IGFs with high affinity, and regulate IGF availability in body fluids and tissues and modulate IGF binding to its receptors. This protein binds IGF-I and IGF-II with relatively low affinity, and belongs to a subfamily of low-affinity IGFBPs. It also stimulates prostacyclin production and cell adhesion. Alternatively spliced transcript variants encoding different isoforms have been described for this gene, and one variant has been associated with retinal arterial macroaneurysm.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IGFBP7
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.1-1.0 mg/mL
Conjugate Biotin
Formulation PBS with no preservative
Gene IGFBP7
Gene Accession No. Q16270
Gene Alias AGM; angiomodulin; FSTL2; IBP7; IBP-7; IGF-binding protein 7; IGFBP; IGFBP rP1; Igfbp7; IGFBP-7; IGFBP-7v; IGFBPRP1; IGFBP-rP1; Insulin like growth factor binding protein; insulin like growth factor binding protein 7; insulin-like growth factor binding protein 7; insulin-like growth factor-binding protein 7; Mac25; MAC25 protein; PGI2-stimulating factor; prostacyclin-stimulating factor; PSF; RAMSVPS; TAF; Tumor-derived adhesion factor
Gene Symbols IGFBP7
Host Species Rabbit
Immunogen E.coli-derived Recombinant Human IGF-BP7.
Purification Method Antigen affinity chromatography
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3490
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.