Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ IGFBPL1 Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA548045
Description
In direct ELISAs, approximately 10% cross-reactivity with recombinant human IGFBP-L1 is observed and less than 1% cross-reactivity with recombinant mouse IGFBP-7 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
IGF-binding proteins prolong the half-life of IGFs and have been shown to either inhibit or stimulate the growth promoting effects of the IGFs in cell culture. They alter the interaction of IGFs with their cell surface receptors. May be a putative tumor suppressor protein.
Specifications
IGFBPL1 | |
Polyclonal | |
Unconjugated | |
Igfbpl1 | |
2810011G06Rik; 2810453O06Rik; bA113O24.1; IGFBP; IGFBPL; Igfbpl1; IGFBP-like protein; IGFBP-related protein 10; Igfbprp4; IGFBP-rP4; Insulin like growth factor binding protein; insulin like growth factor binding protein like 1; insulin like growth factor binding protein-like 1; insulin-like growth factor binding protein related protein 4; insulin-like growth factor binding protein-like 1; insulin-like growth factor-binding protein-like 1; Insulin-like growth factor-binding-related protein 4 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
75426 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q80W15 | |
Igfbpl1 | |
Mouse myeloma cell line NS0-derived recombinant mouse IGFBP-L1 Leu18-Leu270. | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction