Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IGFBPL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567828
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567828 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567828 Supplier Invitrogen™ Supplier No. PA567828
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Predicted to react with Mouse samples.

IGF-binding proteins prolong the half-life of IGFs and have been shown to either inhibit or stimulate the growth promoting effects of the IGFs in cell culture. They alter the interaction of IGFs with their cell surface receptors. May be a putative tumor suppressor protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IGFBPL1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Igfbpl1
Gene Accession No. Q80W15, Q8WX77
Gene Alias 2810011G06Rik; 2810453O06Rik; bA113O24.1; IGFBP; IGFBPL; Igfbpl1; IGFBP-like protein; IGFBP-related protein 10; Igfbprp4; IGFBP-rP4; Insulin like growth factor binding protein; insulin like growth factor binding protein like 1; insulin like growth factor binding protein-like 1; insulin-like growth factor binding protein related protein 4; insulin-like growth factor binding protein-like 1; insulin-like growth factor-binding protein-like 1; Insulin-like growth factor-binding-related protein 4
Gene Symbols Igfbpl1
Host Species Rabbit
Immunogen Synthesized peptide derived from human IGFBPL1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 347252, 75426
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.