Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IGFL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553109
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553109 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553109 Supplier Invitrogen™ Supplier No. PA553109
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TPYLMLCQPH KRCGDKFYDP LQHCCYDDAV VPLARTQTCG NCTFRVCFEQ CCPWTFMVKL INQNCDSART SD Highest antigen sequence identity to the following orthologs: Mouse - 30%, Rat - 33%.

IGFL1 belongs to the insulin-like growth factor family of signaling molecules that play critical roles in cellular energy metabolism and in growth and development, especially prenatal growth.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IGFL1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene IGFL1
Gene Accession No. Q6UW32
Gene Alias APRG644; IGF; IGF like family member 1; IGFL1; IGF-like family member 1; Insulin growth factor-like family member 1; Insulin like growth factor; UNQ644; UNQ644/PRO1274
Gene Symbols IGFL1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human IGFL1. Recombinant protein control fragment (Product #RP-90577).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 374918
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.