Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IGFL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100830
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100830 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100830 Supplier Invitrogen™ Supplier No. PA5100830
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total IGFL1.

IGFL1 belongs to the insulin-like growth factor family of signaling molecules that play critical roles in cellular energy metabolism and in growth and development, especially prenatal growth.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IGFL1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene IGFL1
Gene Accession No. Q6UW32
Gene Alias APRG644; IGF; IGF like family member 1; IGFL1; IGF-like family member 1; Insulin growth factor-like family member 1; Insulin like growth factor; UNQ644; UNQ644/PRO1274
Gene Symbols IGFL1
Host Species Rabbit
Immunogen A synthesized peptide derived from human IGFL1(Accession Q6UW32), corresponding to amino acid residues C21-N71.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 374918
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.