Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IGFL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599820
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599820 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599820 Supplier Invitrogen™ Supplier No. PA599820
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total IGFL2.

The insulin gene family, comprised of insulin, relaxin and insulin-like growth factors I and II (IGF-I and IGF-II), represents a group of structurally related polypeptides whose biological functions have diverged. The IGFs, or somato-medins, constitute a class of polypeptides that have a key role in pre-adolescent mammalian growth. IGF-I and -II are critical regulators of cell proliferation and differentiation. Most of the growth promoting properties of both ligands are mediated by the IGF-I receptor (IGF-IR). IGF- I and -II, respectively known as somatomedin C and somatomedin A, are single chain polypeptides which share an amino acid sequence homology of about 47% with insulin. IGF-I expression is regulated by growth hormone and mediates postnatal growth, while IGF-II is induced by placental lactogen during prenatal development. IGF-II is a fetal growth factor, influenced by placental lactogen and abundantly expressed by placental trophoblasts. IGF-II and IGF-binding protein 1 (IGFBP1) gene variants are associated with overfeeding-induced metabolic changes. The human IGF-II gene maps to chromosome 11p15.5, encoding a 180 amino acid protein which is the precursor to IGF-II.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IGFL2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene IGFL2
Gene Accession No. Q6UWQ7
Gene Alias IGF; IGF like family member 2; IGFL2; IGF-like family member 2; Insulin growth factor-like family member 2; Insulin like growth factor; UNQ645; UNQ645/PRO1275; VPRI645
Gene Symbols IGFL2
Host Species Rabbit
Immunogen A synthesized peptide derived from human IGFL2(Accession Q6UWQ7), corresponding to amino acid residues L18-P68.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 147920
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.