Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IGFL3 Monoclonal Antibody (536825)
GREENER_CHOICE

Catalog No. PIMA524236
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524236 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524236 Supplier Invitrogen™ Supplier No. MA524236
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

In direct ELISAs, no cross-reactivity with recombinant human (rh) IGF-I, rhIGF-II, rhIGFL-1, rhIGFL-4, and recombinant mouse IGFL-3 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.

IGFL3 belongs to the insulin-like growth factor (IGF) family of signaling molecules that play critical roles in cellular energy metabolism and in growth and development, especially prenatal growth.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IGFL3
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 536825
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene IGFL3
Gene Accession No. Q6UXB1
Gene Alias Gm580; IGF; IGF like family member 3; Igfl; Igfl3; IGF-like family member 3; Insulin growth factor-like family member; insulin growth factor-like family member 3; Insulin like growth factor; mIgfl; Rprc483; UNQ483; UNQ483/PRO982
Gene Symbols IGFL3
Host Species Mouse
Immunogen E. coli-derived recombinant human IGFL-3 Thr25-Pro125.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 388555
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.