Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IGFLR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543436
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543436 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543436 Supplier Invitrogen™ Supplier No. PA543436
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: WRCRERPVPA KGHCPLTPGN PGAPSSQERS SPASSIAWRT PEPVPQQAWP Sequence homology: Human: 100%.

TMEM149 is a single-pass type I membrane protein. The exact function of TMEM149 remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IGFLR1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene IGFLR1
Gene Accession No. Q9H665
Gene Alias IGF like family receptor 1; IGF-like family receptor 1; IGFLR1; TMEM149; Transmembrane protein 149; U2 small nuclear RNA auxiliary factor 1-like 4; U2(RNU2) small nuclear RNA auxiliary factor 1-like 4; U2AF1L4
Gene Symbols IGFLR1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human TMEM149 (aa 110-159).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79713
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.