Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IGFLR1/TMEM149 Antibody (905338) [Janelia Fluor™ 646], Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB8296J
Description
IGFLR1/TMEM149 Monoclonal antibody specifically detects IGFLR1/TMEM149 in Human samples. It is validated for Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOFSpecifications
IGFLR1/TMEM149 | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF | |
905338 | |
Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF-ready | |
FLJ22573, transmembrane protein 149, U2 small nuclear RNA auxiliary factor 1-like 4, U2(RNU2) small nuclear RNA auxiliary factor 1-like 4, U2AF1L4 | |
Human embryonic kidney cell line HEK293-derived recombinant human IGFLR1, Met1-Pro163, Accession # Q9H665 | |
0.1 mL | |
Signal Transduction | |
79713 | |
Store at 4°C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction