Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGFLR1/TMEM149 Human anti-Human, Alexa Fluor 488, Clone: Hu14, R&D Systems™

Catalog No. FAB9718G100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB9718G-100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB9718G-100 Supplier R&D Systems Supplier No. FAB9718G100UG

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Human Monoclonal Antibody

IGFLR1/TMEM149 Monoclonal antibody specifically detects IGFLR1/TMEM149 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen IGFLR1/TMEM149
Applications Flow Cytometry
Classification Monoclonal
Clone Hu14
Conjugate Alexa Fluor 488
Dilution Flow Cytometry 0.25 - 1μg/106 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide.
Gene Alias CD221, FLJ22573, IGF1R, IGF-1R, IGFIR, IGF-IR, transmembrane protein 149, U2 small nuclear RNA auxiliary factor 1-like 4, U2(RNU2) small nuclear RNA auxiliary factor 1-like 4, U2AF1L4
Host Species Human
Immunogen Human IGF-I R/IGF1R
Purification Method Protein A or G purified from cell culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 79713
Test Specificity Detects human IGF-I R/IGF1R based on Teprotumumab therapeutic antibody. This non-therapeutic antibody uses the same variable region sequence as the therapeutic antibody Teprotumumab. This product is for research use only.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Supernatant
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.