Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IGFN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571322
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571322 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571322 Supplier Invitrogen™ Supplier No. PA571322
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IGFN1 Polyclonal Antibody for Western Blot

IGFN1 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IGFN1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene IGFN1
Gene Accession No. Q86VF2
Gene Alias eEF1A2 binding protein; EEF1A2-binding protein 1; EEF1A2BP1; IGF; IGFN1; immunoglobulin-like and fibronectin type III domain containing 1; immunoglobulin-like and fibronectin type III domain-containing protein 1; Insulin like growth factor; KY-interacting protein 1; KYIP1
Gene Symbols IGFN1
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human IGFN1.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91156
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.