Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgG Antibody (5A9cc), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP236511F
Description
IgG Monoclonal specifically detects IgG in Human samples. It is validated for ELISA.Specifications
IgG | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
IGHG1 | |
IgG from human serum | |
0.2 mL | |
Adaptive Immunity, Epitope Tags, Immunology | |
3500 | |
Human | |
Purified |
ELISA | |
5A9cc | |
ELISA | |
Immunoglobulin G, ImmunoglobulinG | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Class-specific for human IgG. No allotype restriction. Relatively high affinity. No cross-reactivity with human IgA, IgM, IgE. MAb 5A9 is Fc-region specific. | |
Store at 4C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction