Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgG Antibody (97924) [DyLight 755], Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB110Z
Description
IgG Monoclonal antibody specifically detects IgG in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOFSpecifications
IgG | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry, CyTOF | |
97924 | |
Western Blot, Flow Cytometry, CyTOF-ready | |
Immunoglobulin G, ImmunoglobulinG | |
Mouse myeloma cell line NS0-derived recombinant human IgG1 Fc region | |
0.1 mL | |
Adaptive Immunity, Epitope Tags, Immunology | |
3500 | |
Store at 4°C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction