Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG Antibody (ICO-97), Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$280.00 - $536.00

Specifications

Antigen IgG
Clone ICO-97
Dilution Flow Cytometry 0.5-1ug/million cells, Immunocytochemistry/Immunofluorescence 0.5-1ug/ml, Immunohistochemistry-Frozen 0.5-1ug/ml
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
Classification Monoclonal
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP234251F
SDP
View Documents
Novus Biologicals
NBP2342510.02MG
0.02 mg
Each for $280.00
Only null left
Add to Cart
 
NBP234251A
SDP
View Documents
Novus Biologicals
NBP2342510.1MG
0.1 mg
Each for $536.00
Only null left
Add to Cart
 
NBP234251B
SDP
View Documents
Novus Biologicals
NBP2342510.2MG
0.2 mg N/A N/A N/A
Description

Description

IgG Monoclonal specifically detects IgG in Human samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

IgG
Flow Cytometry 0.5-1ug/million cells, Immunocytochemistry/Immunofluorescence 0.5-1ug/ml, Immunohistochemistry-Frozen 0.5-1ug/ml
Monoclonal
Purified
RUO
Human
P01857
3500
Purified human Ig Gamma Chain
Primary
Store at 4C.
75 kDa
ICO-97
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
Unconjugated
Mouse
Immunology
PBS with 0.05% BSA. with 0.05% Sodium Azide
immunoglobulin heavy constant gamma 1 (G1m marker)
IGHG1
IgG1 κ
Protein A purified
Recognizes a protein of 75kDa, identified as gamma heavy chain of human immunoglobulins. It does not cross-react with alpha (IgA), u (IgM), Epsilon (IgE), or Delta (IgD), heavy chains, T-cells, monocytes, granulocytes, or erythrocytes. This MAb is useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkin's lymphomas. The most common feature of these malignancies is the restricted expression of a single heavy chain class. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.