Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgG Antibody (rIG266), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP279849AF350
Description
IgG Monoclonal specifically detects IgG in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
IgG | |
Monoclonal | |
Alexa Fluor 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
IGHG1 | |
Recombinant full-length human IgG protein | |
0.1 mL | |
Adaptive Immunity, Epitope Tags, Immunology | |
Recognizes a protein of 75kDa, identified as gamma heavy chain of human immunoglobulins. It does not cross-react with alpha (IgA), mu (IgM), epsilon (IgE), or delta (IgD), heavy chains, T-cells, monocytes, granulocytes, or erythrocytes. This monoclonal antibody is useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkins lymphomas. The most common feature of these malignancies is the restricted expression of a single heavy chain class. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant. | |
Store at 4°C in the dark. |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
rIG266 | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
Immunoglobulin G, ImmunoglobulinG | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction