Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgG Antibody (SPM556), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP234428MFV610
Description
IgG Monoclonal antibody specifically detects IgG in Human samples. It is validated for Flow CytometrySpecifications
IgG | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
SPM556 | |
Flow Cytometry | |
Immunoglobulin G, ImmunoglobulinG | |
Purified human IgG | |
0.1 mL | |
Adaptive Immunity, Epitope Tags, Immunology | |
3500 | |
Store at 4C in the dark. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction