Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgG Antibody (SR1919), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | IgG |
---|---|
Clone | SR1919 |
Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 |
Applications | Western Blot, Immunohistochemistry |
Classification | Monoclonal |
Description
IgG Monoclonal antibody specifically detects IgG in Human samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
IgG | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Immunoglobulin G, ImmunoglobulinG | |
A synthesized peptide derived from human IgG (Uniprot #: P01857, P01859, P01860, P01861) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1919 | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Adaptive Immunity, Epitope Tags, Immunology | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
3500 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title