Learn More
IgG Fraction to beta-Galactosidase, Unlabeled, Polyclonal, MP Biomedicals
Shop All MP Biomedicals, Inc ProductsDescription
Specifications
Specifications
| Antigen | IgG Fraction to beta-Galactosidase |
| Applications | ELISA, Fluorescent Immunoassay (FIA), Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Goat |
| Immunogen | Recombinant full length protein of Escherichia coli beta-Galactosidase |
| Quantity | 1 mL |
| Primary or Secondary | Primary |
| Content And Storage | Store unopened product at 2° to 8°C. For long term storage, aliquot and freeze at -20°C. Avoid repeated freeze/thaw cycles. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.