Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG Goat anti-Mouse, Clone: RMG07, Abnova™

Catalog No. 89934571 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-934-571 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-934-571 Supplier Abnova Corporation Supplier No. MAB12777
Add to Cart
Add to Cart

Goat monoclonal antibody raised against full length native mouse IgG.

Specifications

Antigen IgG
Applications ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (PFA fixed), Immunoprecipitation
Classification Monoclonal
Clone RMG07
Conjugate Unconjugated
Description Goat monoclonal antibody raised against full length native mouse IgG.
Dilution ELISA (0.05 ug/mL-1 ug/mL) Flow Cytometry Immunocytochemistry Immunohistochemistry (0.5 ug/mL-2 ug/mL) Immunoprecipitation The optimal working dilution should be determined by the end user.
Formulation In PBS (50% glycerol, 1% BSA, 0.09% sodium azide)
Gene Alias IgG1/Igh-4/VH7183
Gene Symbols Ighg1
Host Species Goat
Immunogen Native purified mouse IgG.
Purification Method Protein G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16017
Test Specificity This antibody reacts to Mouse IgG, including IgG1, IgG2a, IgG2b, and IgG3. No cross reactivity with IgM, IgA, IgE, human IgG, rat IgG, or rabbit IgG.
Target Species Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.