Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG, Goat anti-Rabbit, BV480, Polyclonal, BD Horizon™

Catalog No. BDB564879 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB564879 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB564879 Supplier BD Biosciences Supplier No. 564879
Add to Cart
Add to Cart

Goat Polyclonal Antibody

IgG Polyclonal antibody specifically detects IgG in Rabbit samples. It is validated for Bioimaging.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG
Applications In vitro Assay
Classification Polyclonal
Concentration 0.2mg/mL
Conjugate Brilliant Violet 480
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Goat
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Rabbit
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The polyclonal antibody was purified from antiserum by negative adsorption and affinity chromatography. The antibody was conjugated with BD Horizon BV480 under optimum conditions and unconjugated antibody and free BD Horizon BV480 were removed.
Form Purified
Show More Show Less

For Reseach Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.