Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgG(H+L) Goat anti-Rabbit, Human ads, APC, Polyclonal, Southern Biotech™

Goat Polyclonal Secondary Antibody
Supplier: Southern Biotechnology Associates 405011L
Description
SouthernBiotech is a leading provider of polyclonal secondary antibodies offering high sensitivity and low background for use in a variety of applications.
Specifications
IgG(H+L) | |
Polyclonal | |
APC | |
Goat | |
Affinity Purified | |
RUO | |
Rabbit | |
IgG |
Flow Cytometry, ELISA | |
0.5mg/mL | |
Supplied in PBS/NaN3 and a stabilizing agent | |
Rabbit IgG | |
0.5 mg | |
Secondary | |
Store at 2-8°C; Protect from light; Do not freeze |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction