Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgG(H+L) Goat anti-Rabbit, Human ads, APC, Polyclonal, Southern Biotech™

Goat Polyclonal Secondary Antibody
$532.00 - $688.00
Specifications
| Antigen | IgG(H+L) |
|---|---|
| Concentration | 0.5mg/mL |
| Applications | Flow Cytometry, ELISA |
| Classification | Polyclonal |
| Conjugate | APC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
OB4050-11S
![]() |
Southern Biotechnology Associates
405011S |
0.25 mg |
Each for $532.00
|
|
|||||
OB4050-11L
![]() |
Southern Biotechnology Associates
405011L |
0.5 mg |
Each for $688.00
|
|
|||||
Description
SouthernBiotech is a leading provider of polyclonal secondary antibodies offering high sensitivity and low background for use in a variety of applications.
Specifications
| IgG(H+L) | |
| Flow Cytometry, ELISA | |
| APC | |
| RUO | |
| Supplied in PBS/NaN3 and a stabilizing agent | |
| IgG | |
| Affinity Purified |
| 0.5mg/mL | |
| Polyclonal | |
| Goat | |
| Rabbit | |
| Rabbit IgG | |
| Secondary | |
| Store at 2-8°C; Protect from light; Do not freeze |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title