Learn More
IgG Human, R718, Clone: G18-145, Isotype Control, BD OptiBuild™
Human Monoclonal Antibody
Supplier: BD Biosciences 751917
Description
IgG is an important component of the humoral immune response, helping to control infection. IgG is produced by plasma B-cells and may be found in extracellular fluids, such as blood, lymph, peritoneal, and cerebrospinal fluids. IgG monomers consist of two light and two heavy chains containing two antigen binding sites. There are four IgG subclasses found in human, mouse and rat species, which include IgG1, IgG2, IgG3 and IgG4. The G18-145 monoclonal antibody specifically binds to the heavy chain of human immunoglobulin G subclasses: IgG1, IgG2, IgG3 and IgG4. The G18-145 antibody has been reported not to react with the heavy chains of other human immunoglobulin isotypes.

Specifications
IgG | |
Monoclonal | |
R718 | |
50 μg | |
Isotype Control (Primary) | |
Liquid |
Flow Cytometry | |
G18-145 | |
Human | |
RUO | |
Refrigerate Upon Arrival |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.