Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG, kappa, Hamster, Recombinant, Clone MOPC-21, Isotype Control, Abnova™

Catalog No. 89032418 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-032-418 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-032-418 Supplier Abnova Corporation Supplier No. RAB00075
Add to Cart
Add to Cart

Isotype Control

Specifications

Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone MOPC-21
Conjugate Unconjugated
Dilution Flow Cytometry (1 ug/mL) Negative Control Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (0.02% Proclin 300).
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Isotype Control (Primary)
Test Specificity This antibody of unknown specificity that maybe used as a negative control.
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.