Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgG, kappa, Rabbit, Recombinant, Clone MOPC-21, Isotype Control, Abnova™
Isotype Control
Supplier: Abnova Corporation RAB00114
Specifications
Flow Cytometry, Western Blot | |
MOPC-21 | |
Flow Cytometry (1 ug/mL) Negative Control Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user. | |
200 μg | |
Isotype Control (Primary) | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
IgG κ |
Monoclonal | |
Unconjugated | |
In PBS, pH 7.2 (0.02% Proclin 300). | |
RUO | |
This antibody of unknown specificity that maybe used as a negative control. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction