Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG, kappa, Rabbit, Recombinant, Clone NCRBM/1520R with BSA and Azide, Isotype Control, Abnova™

Catalog No. 89035254 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-254 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-254 Supplier Abnova Corporation Supplier No. RAB00375
Add to Cart
Add to Cart

Isotype Control

Specifications

Applications ELISA, Flow Cytometry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone NCRBM/1520R
Conjugate Unconjugated
Dilution ELISA Flow Cytometry Immunofluorescence Immunohistochemistry Immunoprecipitation Western Blotting The optimal working dilution should be determined by the end user.
Formulation In 10 mM PBS (0.05% BSA, 0.05% sodium azide).
Immunogen Original antibody is raised against recombinant protein corresponding to full length human KRT7.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Isotype Control (Primary)
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.