Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgG Light Chain Antibody (IVA285-1), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP262190APC
Description
IgG Light Chain Monoclonal specifically detects IgG Light Chain in Bovine, Sheep samples. It is validated for Flow Cytometry.Specifications
IgG Light Chain | |
Monoclonal | |
APC | |
PBS with 0.05% Sodium Azide | |
Fraction of Bovine immunoglobulin. | |
0.1 mL | |
Bovine, Sheep | |
IgG1 |
Flow Cytometry | |
IVA285-1 | |
Flow Cytometry | |
Mouse | |
Protein A purified | |
Primary | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction