Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Light Chain Goat anti-Rabbit, HRP, Polyclonal, Secondary Antibody, Novus Biologicals™
SDP

Catalog No. NBP275935 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP275935 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP275935 Supplier Novus Biologicals Supplier No. NBP275935

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Goat Polyclonal Antibody

Light Chain Polyclonal antibody specifically detects Light Chain in Rabbit samples. It is validated for Western Blot.

Specifications

Antigen Light Chain
Applications Western Blot
Classification Polyclonal
Conjugate HRP
Dilution Western Blot 1:10000 - 1:50000
Formulation PBS containing 0.2% BSA with 0.05% Pro-Clean 400
Host Species Goat
Immunogen Rabbit IgG Light Chain antiserum was solid phase adsorbed to ensure specificity.
Purification Method Affinity Purified
Quantity 1 mg
Primary or Secondary Secondary
Test Specificity By immunoelectrophoresis and ELISA this antibody reacts specifically with rabbit light chain. This antibody may cross react with other species.
Target Species Rat, Rabbit
Content And Storage Store at 4°C.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.