Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG light chain (Kappa) Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 501727450 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-7450 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-7450 Supplier Proteintech Group Inc Supplier No. 117811AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunoglobulin classes share the same basic four polypeptide chain structure of two heavy chains (five types) and two light chains (kappa, lambda; both having a molecular weight of 22.5 kDa). Kappa and lambda consist of a variable region and a constant region and can easily be differentiated by the antigenic properties of the constant region. The ratio of kappa to lambda is 70:30. Antibodies to the kappa light chain are reportedly useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkin′s lymphomas. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore maligt.

Specifications

Antigen IgG light chain (Kappa)
Applications Western Blot
Classification Polyclonal
Concentration 0.32 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene IGKV1-5
Gene Accession No. P01602
Gene Alias IGKV, IGKV1 5, IGKV15, IGKV1-5, L12, L12a, V1
Gene Symbols IGKV1-5
Host Species Rabbit
Immunogen IgG light chain (Kappa) Fusion Protein Ag2406
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 28299
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.