Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgG light chain (lambda) Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 115411AP150UL
Description
Immunoglobulin classes share the same basic four polypeptide chain structure of two heavy chains (five types) and two light chains (kappa, lambda; both having a molecular weight of 22.5 kDa). Kappa and lambda consist of a variable region and a constant region and can easily be differentiated by the antigenic properties of the constant region. The ratio of kappa to lambda is 70:30. Antibodies to the kappa light chain are reportedly useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkin′s lymphomas. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore maligt.Specifications
| IgG light chain (lambda) | |
| Polyclonal | |
| Unconjugated | |
| IGL | |
| IGL | |
| IgG light chain (lambda) Fusion Protein Ag2064 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| IGL, IGL-, immunoglobulin lambda locus | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 3535 | |
| -20°C | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction