Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgG Mouse anti-Human, RB545, Clone: G18-145, BD Horizon™
Mouse Monoclonal Antibody
$162.00 - $396.00
Specifications
Antigen | IgG |
---|---|
Clone | G18-145 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | RB545 |
Description
IgG Monoclonal antibody specifically detects IgG in Human samples. It is validated for Flow cytometrySpecifications
IgG | |
Flow Cytometry | |
RB545 | |
Mouse | |
Human | |
IGHG1, IGHG2, IGHG3 and IGHG4 | |
Primary | |
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unreacted dye was removed. Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
G18-145 | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
IgG1 κ | |
Affinity Chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title