Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IGG Rabbit anti-Mouse, Clone: RM104, Abnova™
Rabbit monoclonal antibody raised against native mouse IgG.
Supplier: Abnova Corporation MAB12765
Specifications
IGG | |
Monoclonal | |
Unconjugated | |
ELISA (0.005 - 0.2 ug/mL) Flow Cytometry Immunocytochemistry (0.5 - 2 ug/mL) Immunohistochemistry (0.5 - 2 ug/mL) Immunoprecipitation Western Blot (0.1 - 0.5 ug/mL) The optimal working dilution should be determined by the end user. | |
IgG1/Igh-4/VH7183 | |
Rabbit | |
Protein A affinity purification | |
RUO | |
16017 | |
Mouse | |
Liquid |
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
RM104 | |
Rabbit monoclonal antibody raised against native mouse IgG. | |
In PBS (50% glycerol, 1% BSA, 0.09% sodium azide) | |
Ighg1 | |
Native purified IgG from mouse serum. | |
100 μg | |
Primary | |
This antibody reacts to the Fc region of all subclasses of mouse IgG. No cross reactivity with mouse IgM, IgA, IgE, human IgG, and rat IgG. It may cross react to goat IgG. | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction