Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit IgG Isotype Control, Allophycocyanin/Cy7, Novus Biologicals™
SDP

Catalog No. NB001388 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB001388 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB001388 Supplier Novus Biologicals Supplier No. NBP224891APCCY7
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IgG Polyclonal specifically detects IgG in Rabbit samples. It is validated for Chromatin Immunoprecipitation, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, In vitro assay.

Specifications

Antigen IgG
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), In vitro Assay, ChIP Assay
Classification Polyclonal
Conjugate APC-Cyanine7
Dilution Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, In vitro assay, Chromatin Immunoprecipitation (ChIP)
Formulation PBS with 0.05% Sodium Azide
Gene Alias Immunoglobulin G, ImmunoglobulinG
Gene Symbols IGHG1
Host Species Rabbit
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Isotype Control (Primary)
Gene ID (Entrez) 3500
Target Species Rabbit
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.