Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG, Rat, Polyclonal, Isotype Control, Novus Biologicals™
SDP

Catalog No. NBP22195125 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.25 mg
0.5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP22195125 0.25 mg
NBP22195105 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP22195125 Supplier Novus Biologicals Supplier No. NBP2219510.25MG

Rat Polyclonal Antibody

IgG Polyclonal specifically detects IgG in Rat samples. It is validated for .

Specifications

Antigen IgG
Classification Polyclonal
Conjugate Unconjugated
Formulation 0.2 um filtered solution in PBS.
Host Species Rat
Purification Method Protein A or G purified
Quantity 0.25 mg
Primary or Secondary Isotype Control (Primary)
Reconstitution Reconstitute with 1.0 mL PBS.
Target Species Rat
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.